5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile 97%

Warning

H27358-03 H27358-06
AAH27358-03EA 59.84 USD
AAH27358-03 AAH27358-06
5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile 97%
5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
Formula: C₁₀H₇ClN₄
MW: 218.65 g/mol
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00052031
CAS Number: 51516-67-7
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR