Dimethyl isopropenyl phosphate 95%

H27353-14 H27353-22 H27353-06
AAH27353-22EA 236.89 USD
AAH27353-22 AAH27353-14 AAH27353-06
Dimethyl isopropenyl phosphate 95%
Dimethyl isopropenyl phosphate
Formula: C₅H₁₁O₄P
MW: 166.11 g/mol
Boiling Pt: 120 °C/7.5 mm
Storage Temperature: Ambient
MDL Number: MFCD09953485
CAS Number: 4185-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR