(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol(S)-BIPHEN H2(S)-(-)-5,5',6,6'-Tetramethyl-3,3'-di-t-butyl-1,1'-biphenyl-2,2'-diol

H27342-MD H27342-03
AAH27342-MDEA 165.99 USD
AAH27342-MD AAH27342-03
(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol 99%
(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol
Formula: C₂₄H₃₄O₂
MW: 354.526 g/mol
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD01862439
CAS Number: 205927-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR