9,9-Dimethylxanthene ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 9,9-Dimethyl-9H-xanthene

H27340-06
AAH27340-06EA 131.99 USD
AAH27340-06
9,9-Dimethylxanthene ≥98%
9,9-Dimethylxanthene
Formula: C₁₅H₁₄O
MW: 210.28 g/mol
Boiling Pt: 114…115 °C/0.6 mm
Melting Pt: 40…43 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134434
CAS Number: 19814-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR