(2-Methylcyclopropyl)methanol cis- and trans- mixture 98%

Warning

H27337-03 H27337-09
AAH27337-03EA 37.12 USD
AAH27337-03 AAH27337-09
(2-Methylcyclopropyl)methanol cis- and trans- mixture 98%
(2-Methylcyclopropyl)methanol
Formula: C₅H₁₀O
Boiling Pt: 133 °C
Density: 0.87
Flash Pt: 39 °C(102 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009683
CAS Number: 6077-72-1
UN: 1987
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR