4-Chloro-2-(5-isoxazolyl)phenol 97%

Warning

H27333-03 H27333-06
AAH27333-06EA 71.57 USD
AAH27333-06 AAH27333-03
4-Chloro-2-(5-isoxazolyl)phenol 97%
4-Chloro-2-(5-isoxazolyl)phenol
Formula: C₉H₆ClNO₂
Melting Pt: 200…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00218660
CAS Number: 86176-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR