1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥99%

Synonyms: 1-Butyl-4-((4-methoxyphenyl)ethynyl)benzene

H27304-03 H27304-09
AAH27304-09EA 287.34 USD
AAH27304-09 AAH27304-03
1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥99%
1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene
Formula: C₁₉H₂₀O
MW: 264.37 g/mol
Boiling Pt: 172…172 °C/1 mm
Melting Pt: 40 °C (C-Nematic)
Storage Temperature: Ambient
MDL Number: MFCD04038796
CAS Number: 35684-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR