4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester 95%
|
Warning
|
H27292-03
H27292-MD
AAH27292-MDEA
269.72
USD
AAH27292-MD
AAH27292-03
4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester 95%
4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester
Formula:
C₁₇H₂₀BClN₂O₃ Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD06798267 CAS Number: 1073354-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...