4-Cyano-4-(4-fluorophenyl)cyclohexanone 98%
|
Warning
|
Synonyms:
1-(4-Fluorophenyl)-4-oxocyclohexane-1-carbonitrile
H27285-03
H27285-MD
AAH27285-03EA
194.25
USD
AAH27285-03
AAH27285-MD
4-Cyano-4-(4-fluorophenyl)cyclohexanone 98%
4-Cyano-4-(4-fluorophenyl)cyclohexanone
Formula:
C₁₃H₁₂FNO MW: 217.25 Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044815 EINECS: 260-111-7 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...