3-(2-Furyl)propanoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Furan-2-yl)propanoic acid
H27273-06
AAH27273-06EA
127.99
USD
AAH27273-06
3-(2-Furyl)propanoic acid 97%
3-(2-Furyl)propanoic acid
Formula:
C₇H₈O₃ MW: 140.138 g/mol Melting Pt: 56…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005346 CAS Number: 935-13-7 EINECS: 213-298-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...