1-Aminocyclopropanecarboxylic acid hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-azanylcyclopropane-1-carboxylic acid hydrochloride1-aminocyclopropane-1-carboxylic acid hydrochlorideACPCCyclopropanecarboxylic acid, 1-amino-, hydrochloride

H27264-MD H27264-03
AAH27264-03EA 405.99 USD
AAH27264-03 AAH27264-MD
1-Aminocyclopropanecarboxylic acid hydrochloride 97%
1-Aminocyclopropanecarboxylic acid hydrochloride
Formula: C₄H₈ClNO₂
MW: 137.57 g/mol
Melting Pt: ca 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012545
CAS Number: 68781-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR