3-(Boc-amino)phenylboronic acid 95%
|
Warning
|
Synonyms:
(3-((tert-Butoxycarbonyl)amino)phenyl)boronic acid
, 3-(Boc-amino)benzeneboronic acid
H27258-06
H27258-03
AAH27258-06EA
272.95
USD
AAH27258-06
AAH27258-03
3-(Boc-amino)phenylboronic acid 95%
3-(Boc-amino)phenylboronic acid
Formula:
C₁₁H₁₆BNO₄ MW: 237.06 g/mol Melting Pt: ca 155 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03411945 CAS Number: 380430-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...