Pyridazine-4-carboxylic acid 97%
|
Warning
|
Synonyms:
4-Pyridazinecarboxylic acid
H27246-06
H27246-03
AAH27246-03EA
103.99
USD
AAH27246-03
AAH27246-06
Pyridazine-4-carboxylic acid 97%
Pyridazine-4-carboxylic acid
Formula:
C₅H₄N₂O₂ MW: 124.10 g/mol Melting Pt: ca 244 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011576 CAS Number: 50681-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...