Ethyl (E)-2-cyano-3-ethoxycrotonate 98%

Supplier: Thermo Scientific Chemicals
Warning

H27244-14 H27244-06
AAH27244-06EA 41.59 USD
AAH27244-06 AAH27244-14
Ethyl (E)-2-cyano-3-ethoxycrotonate 98%
Ethyl (E)-2-cyano-3-ethoxycrotonate
Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD01743485
CAS Number: 932750-29-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR