(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: cis-4-Fluoro-L-prolinamide hydrochloride

H27235-06 H27235-03
AAH27235-03EA 219.99 USD
AAH27235-03 AAH27235-06
(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride 98%
(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride
Formula: C₅H₁₀ClFN₂O
MW: 168.6 g/mol
Melting Pt: 231…236 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD07368567
CAS Number: 426844-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR