Methyl-7-bromoheptanoate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl 7-bromoheptanoate
H27234-06
H27234-14
AAH27234-14EA
277.99
USD
AAH27234-14
AAH27234-06
Methyl-7-bromoheptanoate 98%
Methyl-7-bromoheptanoate
Formula:
C₈H₁₅BrO₂ MW: 223.12 g/mol Density: 1.257 Storage Temperature: Ambient |
MDL Number:
MFCD02258672 CAS Number: 54049-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...