2-(1-Naphthalene)ethanol 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-(1-Naphthyl)ethanol
, 2-(Naphthalen-1-yl)ethanol
H27218-09
H27218-18
AAH27218-09EA
82.89
USD
AAH27218-09
AAH27218-18
2-(1-Naphthalene)ethanol 95%
2-(1-Naphthalene)ethanol
Formula:
C₁₂H₁₂O MW: 172.23 g/mol Boiling Pt: 186 °C/17 mm Melting Pt: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004050 CAS Number: 773-99-9 EINECS: 212-260-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...