2,2,2-Trifluoroethyl formate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H27212-03
H27212-06
AAH27212-06EA
143
USD
AAH27212-06
AAH27212-03
2,2,2-Trifluoroethyl formate 95%
2,2,2-Trifluoroethyl formate
Formula:
C₃H₃F₃O₂ Boiling Pt: 59…60 °C Density: 1.317 Flash Pt: 7 °C(45 °F) Storage Temperature: Ambient |
UN:
1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...