6-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile 97%
|
Warning
|
H27198-03
AAH27198-03EA
43.21
USD
AAH27198-03
6-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile 97%
6-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile
Formula:
C₁₂H₁₂N₂O₂ MW: 216.24 g/mol Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427105 CAS Number: 351003-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...