1,2-Bis(3-indenyl)ethane 98%

Synonyms: 1,2-Di(1H-inden-3-yl)ethane3,3'-Ethylenebis(1H-indene)

H27195-06
AAH27195-06EA 91.37 USD
AAH27195-06
1,2-Bis(3-indenyl)ethane 98%
1,2-Bis(3-indenyl)ethane
Formula: C₂₀H₁₈
MW: 258.36 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00191814
CAS Number: 18657-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR