3,5-Dibromo-4-cyanopyridine 97%

Warning

Synonyms: 3,5-Dibromoisonicotinonitrile

H27186-03 H27186-06 H27186-MD
AAH27186-MDEA 72.37 USD
AAH27186-MD AAH27186-06 AAH27186-03
3,5-Dibromo-4-cyanopyridine 97%
3,5-Dibromo-4-cyanopyridine
Formula: C₆H₂Br₂N₂
MW: 261.90 g/mol
Melting Pt: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD09263321
CAS Number: 870244-34-1
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR