6-Nitrochromone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Nitro-4H-chromen-4-one

H27167-03 H27167-06 H27167-14
AAH27167-06EA 74.79 USD
AAH27167-06 AAH27167-03 AAH27167-14
6-Nitrochromone 97%
6-Nitrochromone
Formula: C₉H₅NO₄
MW: 191.15 g/mol
Melting Pt: 172…175 °C
Storage Temperature: Ambient
MDL Number: MFCD02954226
CAS Number: 51484-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR