5-Pyrimidineboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(5-Pyrimidyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

H27147-MD H27147-03
AAH27147-03EA 225.99 USD
AAH27147-03 AAH27147-MD
5-Pyrimidineboronic acid pinacol ester 95%
5-Pyrimidineboronic acid pinacol ester
Formula: C₁₀H₁₅BN₂O₂
MW: 206.05 g/mol
Melting Pt: 60…66 °C
Storage Temperature: Ambient
MDL Number: MFCD05155223
CAS Number: 321724-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR