2,5-Dichloro-3-methylthiophene 96%
|
Warning
|
H27129-03
AAH27129-03EA
45.48
USD
AAH27129-03
2,5-Dichloro-3-methylthiophene 96%
2,5-Dichloro-3-methylthiophene
Formula:
C₅H₄Cl₂S Density: 1.331 Flash Pt: 77 °C(171 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00022490 CAS Number: 17249-90-0 UN: 2810 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...