1-(1-Boc-3-azetidinyl)pyrrolidine 94%

Warning

H27111-MD H27111-03
AAH27111-03EA 272.07 USD
AAH27111-03 AAH27111-MD
1-(1-Boc-3-azetidinyl)pyrrolidine 94%
1-(1-Boc-3-azetidinyl)pyrrolidine
Formula: C₁₂H₂₂N₂O₂
MW: 226.32
Storage Temperature: Ambient
MDL Number: MFCD09860616
CAS Number: 1019008-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR