3-Methyl-D-phenylalanine 98%

Synonyms: (R)-2-Amino-3-(m-tolyl)propanoic acid

H27108-MD
AAH27108-MDEA 103.56 USD
AAH27108-MD
3-Methyl-D-phenylalanine 98%
3-Methyl-D-phenylalanine
Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01860871
CAS Number: 114926-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR