4-Chloro-3-nitrophenylboronic acid 97%
|
Warning
|
Synonyms:
4-Chloro-3-nitrobenzeneboronic acid
H27093-MC
AAH27093-MCEA
83.49
USD
AAH27093-MC
4-Chloro-3-nitrophenylboronic acid 97%
4-Chloro-3-nitrophenylboronic acid
Formula:
C₆H₅BClNO₄ MW: 201.37 g/mol Melting Pt: ca 275 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02258950 CAS Number: 151169-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...