Cyclopentylboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27081-09
H27081-03
AAH27081-09EA
266.99
USD
AAH27081-09
AAH27081-03
Cyclopentylboronic acid 95%
Cyclopentylboronic acid
Formula:
C₅H₁₁BO₂ MW: 113.95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01074541 CAS Number: 63076-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...