5-Bromo-2-hydroxy-4,6-dimethylnicotinonitrile 98%

Warning

Synonyms: 5-Bromo-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

H27058-03 H27058-MD
AAH27058-MDEA 94.86 USD
AAH27058-MD AAH27058-03
5-Bromo-2-hydroxy-4,6-dimethylnicotinonitrile 98%
5-Bromo-2-hydroxy-4,6-dimethylnicotinonitrile
Formula: C₈H₇BrN₂O
MW: 227.07 g/mol
Melting Pt: ca 256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00052651
CAS Number: 23819-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR