(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2S)-1-[(tert-butoxy)carbonyl]-4,4-difluoropyrrolidine-2-carboxylic acid
, N-(tert-Butoxycarbonyl)-4,4-difluoro-L-proline
, N-Boc-4,4-difluoro-L-proline
, (S)-1-Boc-4,4-difluoro-2-pyrrolidinecarboxylic Acid
, (S)-1-(tert-Butoxycarbonyl)-4,4-difluoropyrrolidine-2-carboxylic acid
H27052-MD
H27052-03
AAH27052-03EA
265.99
USD
AAH27052-03
AAH27052-MD
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97%
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid
Formula:
C₁₀H₁₅F₂NO₄ MW: 251.23 g/mol Melting Pt: 120…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094917 CAS Number: 203866-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...