2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester 95%

H27051-06 H27051-14 H27051-03
AAH27051-14EA 502.83 USD
AAH27051-14 AAH27051-03 AAH27051-06
2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester 95%
2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester
Formula: C₁₈H₂₉BO₂
Storage Temperature: Ambient
MDL Number: MFCD09953477
CAS Number: 1073355-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR