N-Acetyl-D-leucine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ac-D-Leu-OH
H27046-06
H27046-03
H27046-14
AAH27046-06EA
276.99
USD
AAH27046-06
AAH27046-03
AAH27046-14
N-Acetyl-D-leucine 99%
N-Acetyl-D-leucine
Formula:
C₈H₁₅NO₃ MW: 173.2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00066069 CAS Number: 19764-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...