1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazole-5-boronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole

H27043-03 H27043-MD
AAH27043-MDEA 196.47 USD
AAH27043-MD AAH27043-03
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazole-5-boronic acid pinacol ester 95%
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazole-5-boronic acid pinacol ester
Formula: C₁₅H₂₉BN₂O₃Si
MW: 324.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037502
CAS Number: 903550-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR