3-(Boc-amino)phenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-(Boc-amino)benzeneboronic acid pinacol ester
, tert-Butyl (3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate
H27026-06
H27026-03
AAH27026-03EA
92.89
USD
AAH27026-03
AAH27026-06
3-(Boc-amino)phenylboronic acid pinacol ester 97%
3-(Boc-amino)phenylboronic acid pinacol ester
Formula:
C₁₇H₂₆BNO₄ MW: 319.21 g/mol Melting Pt: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD03789256 CAS Number: 330793-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...