5-Amino-2-mercaptobenzimidazole 96%

Warning

Synonyms: 5-Amino-1H-benzoimidazole-2-thiol

H26999-14 H26999-22
AAH26999-14EA 81.15 USD
AAH26999-14 AAH26999-22
5-Amino-2-mercaptobenzimidazole 96%
5-Amino-2-mercaptobenzimidazole
Formula: C₇H₇N₃S
MW: 165.22 g/mol
Melting Pt: 240…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00022671
CAS Number: 2818-66-8
EINECS: 220-574-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR