6-Chloro-7-methylchromone 98%
|
Warning
|
Synonyms:
6-Chloro-7-methyl-4H-chromen-4-one
H26997-03
AAH26997-03EA
102.46
USD
AAH26997-03
6-Chloro-7-methylchromone 98%
6-Chloro-7-methylchromone
Formula:
C₁₀H₇ClO₂ MW: 194.62 g/mol Melting Pt: 169…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239401 CAS Number: 67029-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...