Dicyclohexylphenylphosphine 98%

Warning

Synonyms: Dicyclohexyl(phenyl)phosphine

H26992-06 H26992-03
AAH26992-03EA 77.98 USD
AAH26992-03 AAH26992-06
Dicyclohexylphenylphosphine 98%
Dicyclohexylphenylphosphine
Formula: C₁₈H₂₇P
MW: 274.38 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00003854
CAS Number: 6476-37-5
EINECS: 229-334-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR