2,3,6,7-Tetrahydro-(1H)-1,4-diazepin-5-(4H)-one 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Homopiperazin-5-one

H26990-MD H26990-03
AAH26990-03EA 329.99 USD
AAH26990-03 AAH26990-MD
2,3,6,7-Tetrahydro-(1H)-1,4-diazepin-5-(4H)-one 95%
2,3,6,7-Tetrahydro-(1H)-1,4-diazepin-5-(4H)-one
Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00766804
CAS Number: 34376-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR