4-Formyl-3-(trifluoromethyl)phenylboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Formyl-3-(trifluoromethyl)benzeneboronic acid
H26988-03
AAH26988-03EA
515
USD
AAH26988-03
4-Formyl-3-(trifluoromethyl)phenylboronic acid 95%
4-Formyl-3-(trifluoromethyl)phenylboronic acid
Formula:
C₈H₆BF₃O₃ Melting Pt: 200…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037498 CAS Number: 1072944-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...