2,6-Dimethoxy-4-methylbenzaldehyde 97%
|
Warning
|
H26982-09
H26982-03
AAH26982-03EA
52.84
USD
AAH26982-03
AAH26982-09
2,6-Dimethoxy-4-methylbenzaldehyde 97%
2,6-Dimethoxy-4-methylbenzaldehyde
Formula:
C₁₀H₁₂O₃ Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD01321089 CAS Number: 6937-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...