2,6-Dichloro-4-nitropyridine-N-oxide 97%

Warning

H26979-06
AAH26979-06EA 54.84 USD
AAH26979-06
2,6-Dichloro-4-nitropyridine-N-oxide 97%
2,6-Dichloro-4-nitropyridine N-oxide
Formula: C₅H₂Cl₂N₂O₃
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00955611
CAS Number: 2587-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR