2-Benzyloxy-3-pyridineboronic acid 95%
|
Warning
|
H26978-03
H26978-MD
AAH26978-MDEA
119.88
USD
AAH26978-MD
AAH26978-03
2-Benzyloxy-3-pyridineboronic acid 95%
2-Benzyloxy-3-pyridineboronic acid
Formula:
C₁₂H₁₂BNO₃ Melting Pt: 145…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD06798264 CAS Number: 1072952-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...