(S)-(-)-XylBINAP 98%

Warning

Synonyms: (S)-2,2'-Bis(bis(3,5-dimethylphenyl)phosphino)-1,1'-binaphthalene(S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl

H26970-MC H26970-ME
AAH26970-MEEA 356.51 USD
AAH26970-ME AAH26970-MC
(S)-(-)-XylBINAP 98%
(S)-(-)-XylBINAP
Formula: C₅₂H₄₈P₂
MW: 734.90 g/mol
Melting Pt: 201…203 °C
Storage Temperature: Ambient
MDL Number: MFCD01630821
CAS Number: 135139-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR