Bis(cyclopentadienyl)dimethylhafnium 97+%
Synonyms:
Bis(cyclopentadienyl)dimethylhafnium(IV)
H26962-06
H26962-03
AAH26962-03EA
98.05
USD
AAH26962-03
AAH26962-06
Bis(cyclopentadienyl)dimethylhafnium 97+%
Bis(cyclopentadienyl)dimethylhafnium
Formula:
C₁₂H₁₆Hf Melting Pt: ca 118 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02093015 CAS Number: 37260-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...