5-Bromo-2-(2-furyl)pyridine 95%

Warning

H26942-MD H26942-03
AAH26942-MDEA 158.73 USD
AAH26942-MD AAH26942-03
5-Bromo-2-(2-furyl)pyridine 95%
5-Bromo-2-(2-furyl)pyridine
Formula: C₉H₆BrNO
MW: 224.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037418
CAS Number: 942206-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR