3-(Methylsulfinyl)phenylacetic acid 96%

Warning

H26921-06 H26921-03
AAH26921-03EA 104.67 USD
AAH26921-03 AAH26921-06
3-(Methylsulfinyl)phenylacetic acid 96%
3-(Methylsulfinyl)phenylacetic acid
Formula: C₉H₁₀O₃S
MW: 198.24 g/mol
Melting Pt: 110…112°C
Storage Temperature: Ambient
MDL Number: MFCD09953474
CAS Number: 1027730-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR