(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-Dap-OH
, Boc-Dpr-OH
, 3-Amino-N-Boc-L-alanine
, 3-Amino-N-(tert-butoxycarbonyl)-L-alanine
, Nalpha-Boc-2,3-diaminopropionic Acid
H26919-03
AAH26919-03EA
74.69
USD
AAH26919-03
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 97%
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid
Formula:
C₈H₁₆N₂O₄ MW: 204.23 g/mol Melting Pt: ca 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00236843 CAS Number: 73259-81-1 |
Specification Test Results
Optical rotation | -14° (c=1 in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...