1-Ethyl-4-[(4-n-hexylphenyl)ethynyl]benzene ≥99%

H26908-03
AAH26908-03EA 39.77 USD
AAH26908-03
1-Ethyl-4-[(4-n-hexylphenyl)ethynyl]benzene ≥99%
1-Ethyl-4-[(4-n-hexylphenyl)ethynyl]benzene
Formula: C₂₂H₂₆
Boiling Pt: 172…174 °C/0.5 mm
Melting Pt: 10…12 °C
Storage Temperature: Ambient
MDL Number: MFCD09909569
CAS Number: 117923-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR