Di-tert-butylphenylphosphonium tetrafluoroborate 99%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Di-tert-butyl(phenyl)phosphonium tetrafluoroborate

H26904-03
AAH26904-03EA 284 USD
AAH26904-03
Di-tert-butylphenylphosphonium tetrafluoroborate 99%
Di-tert-butylphenylphosphonium tetrafluoroborate
Formula: C₁₄H₂₄BF₄P
MW: 310.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08704553
CAS Number: 612088-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR