1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Ethyl-4-[(4-methylphenyl)ethynyl]benzene1-(4-Ethylphenyl)-2-(p-tolyl)acetylene1-Ethyl-4-[(p-tolyl)ethynyl]benzene

H26882-03
AAH26882-03EA 58.69 USD
AAH26882-03
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥99%
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene
Formula: C₁₇H₁₆
MW: 220.315 g/mol
Boiling Pt: 135…136 °C/1 mm
Melting Pt: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD04038793
CAS Number: 22692-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR